MAT047023449 Matrix Scientific

3-Bromopropyl isothiocyanate

  • MDL number MFCD00060421
  • Cas Number : 2799-73-7
  • Molecular Weight : 180.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.54
Country of Origin USA
Boiling point 126-128�mm
Product Link http://www.matrixscientific.com/004962.html

Safety Information

hcodes Toxic/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/004962.html
Availability details for
Quantity
  • Availability
  • Add to Cart