MAT047023457 Matrix Scientific

2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride

  • MDL number MFCD01631474
  • Cas Number : 143096-86-0
  • Molecular Weight : 220.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/004974.html

Safety Information

hcodes Corrosive/Stored under Argon/Moisture Sensitive
MSDS http://www.matrixscientific.com/004974.html
Availability details for
Quantity
  • Availability
  • Add to Cart