MAT047023463 Matrix Scientific

1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether

  • MDL number MFCD01320775
  • Cas Number : 32778-07-7
  • Molecular Weight : 218.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Ambient
Country of Origin USA
Boiling point 60-61
Product Link http://www.matrixscientific.com/004979.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004979.html
Availability details for
Quantity
  • Availability
  • Add to Cart