MAT047023471 Matrix Scientific

1,1-Dichloro-2-iodo-1,2,2-trifluoroethane

  • MDL number MFCD02093303
  • Cas Number : 661-66-5
  • Molecular Weight : 278.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 70
Density 2.189/25
Country of Origin USA
Boiling point 53�0 Torr
Product Link http://www.matrixscientific.com/004996.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004996.html
Availability details for
Quantity
  • Availability
  • Add to Cart