MAT047023474 Matrix Scientific

4,5-Difluoro-2-methylbenzimidazole

  • MDL number MFCD00665871
  • Cas Number : 229957-09-9
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250
Country of Origin USA
Product Link http://www.matrixscientific.com/004999.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004999.html
Availability details for
Quantity
  • Availability
  • Add to Cart