MAT047023477 Matrix Scientific

1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)-propane

  • MDL number MFCD01862044
  • Cas Number : 138690-25-2
  • Molecular Weight : 234.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 78-79
Product Link http://www.matrixscientific.com/005003.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005003.html
Availability details for
Quantity
  • Availability
  • Add to Cart