MAT047023478 Matrix Scientific

2,4-Dichloro-3,5-dinitrobenzotrifluoride

  • MDL number MFCD00042480
  • Cas Number : 29091-09-6
  • Molecular Weight : 305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-78
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/005005.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/005005.html
Availability details for
Quantity
  • Availability
  • Add to Cart