MAT047023483 Matrix Scientific

2,6-Difluoro-3-methylanisole

  • MDL number MFCD01631341
  • Cas Number : 261763-33-1
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 53�mm
Product Link http://www.matrixscientific.com/005012.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005012.html
Availability details for
Quantity
  • Availability
  • Add to Cart