MAT047023490 Matrix Scientific

2,6-Difluoro-3-methylbenzyl bromide

  • MDL number MFCD01631336
  • Cas Number : 261763-44-4
  • Molecular Weight : 221.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 76�Torr
Product Link http://www.matrixscientific.com/005019.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/005019.html
Availability details for
Quantity
  • Availability
  • Add to Cart