MAT047023491 Matrix Scientific

2,6-Difluoro-3-methylphenol

  • MDL number MFCD01631340
  • Cas Number : 261763-46-6
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 182
Product Link http://www.matrixscientific.com/005020.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005020.html
Availability details for
Quantity
  • Availability
  • Add to Cart