MAT047023504 Matrix Scientific

Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester

  • MDL number MFCD00054670
  • Cas Number : 133609-46-8
  • Molecular Weight : 842.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.74
Country of Origin USA
Boiling point 70-80�1mm
Product Link http://www.matrixscientific.com/001416.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001416.html
Availability details for
Quantity
  • Availability
  • Add to Cart