MAT047023539 Matrix Scientific

2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane

  • MDL number MFCD00054715
  • Cas Number : 3330-16-3
  • Molecular Weight : 618.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -106.7
Purity 95
Density 1.72
Country of Origin USA
Boiling point 151-154
Product Link http://www.matrixscientific.com/005085.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005085.html
Availability details for
Quantity
  • Availability
  • Add to Cart