MAT047023541 Matrix Scientific

2,4-Dichloro-5-fluorotoluene

  • MDL number MFCD06246852
  • Cas Number : 86522-86-3
  • Molecular Weight : 179.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/005102.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005102.html
Availability details for
Quantity
  • Availability
  • Add to Cart