MAT047023544 Matrix Scientific

2,4-Dichloro-5-fluorobenzamide

  • MDL number MFCD01631478
  • Cas Number : 135748-33-3
  • Molecular Weight : 208.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-160
Country of Origin USA
Product Link http://www.matrixscientific.com/005105.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005105.html
Availability details for
Quantity
  • Availability
  • Add to Cart