MAT047023558 Matrix Scientific

3,4-Dichloro-5-nitrobenzotrifluoride

  • MDL number MFCD01862035
  • Cas Number : 657-02-3
  • Molecular Weight : 260

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 115-118� Torr
Product Link http://www.matrixscientific.com/005120.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/005120.html
Availability details for
Quantity
  • Availability
  • Add to Cart