MAT047023560 Matrix Scientific

2,4-Dichloro-6-(trifluoromethyl)phenylhydrazine

  • MDL number MFCD00174090
  • Cas Number : 107047-29-0
  • Molecular Weight : 245.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-43
Country of Origin USA
Product Link http://www.matrixscientific.com/005121.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005121.html
Availability details for
Quantity
  • Availability
  • Add to Cart