MAT047023565 Matrix Scientific

2,3-Difluoro-6-nitroaniline

  • MDL number MFCD00233313
  • Cas Number : 211693-73-1
  • Molecular Weight : 174.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Country of Origin USA
Product Link http://www.matrixscientific.com/005126.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005126.html
Availability details for
Quantity
  • Availability
  • Add to Cart