MAT047023566 Matrix Scientific

3,4-Dichloro-6-nitrobenzotrifluoride

  • MDL number MFCD00236666
  • Cas Number : 50594-31-5
  • Molecular Weight : 260

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005127.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005127.html
Availability details for
Quantity
  • Availability
  • Add to Cart