MAT047023602 Matrix Scientific

2,5-Difluorobenzamide

  • MDL number MFCD00015548
  • Cas Number : 85118-03-2
  • Molecular Weight : 157.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/005167.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005167.html
Availability details for
Quantity
  • Availability
  • Add to Cart