MAT047023627 Matrix Scientific

3,4-Difluorobenzhydrazide

  • MDL number MFCD00672992
  • Cas Number : 229957-07-7
  • Molecular Weight : 172.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Country of Origin USA
Product Link http://www.matrixscientific.com/005189.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005189.html
Availability details for
Quantity
  • Availability
  • Add to Cart