MAT047023650 Matrix Scientific

3,5-Difluorobenzophenone

  • MDL number MFCD00061140
  • Cas Number : 179113-89-4
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-59
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005211.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005211.html
Availability details for
Quantity
  • Availability
  • Add to Cart