MAT047023660 Matrix Scientific

3,4-Difluorobenzotrifluoride

  • MDL number MFCD00077510
  • Cas Number : 32137-19-2
  • Molecular Weight : 182.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-98
Country of Origin USA
Boiling point 104
Product Link http://www.matrixscientific.com/005220.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/005220.html
Availability details for
Quantity
  • Availability
  • Add to Cart