MAT047023665 Matrix Scientific

3-(3,4-Dichlorophenyl)-3-oxopropanenitrile

  • MDL number MFCD00067919
  • Cas Number : 4640-68-0
  • Molecular Weight : 214.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/005225.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005225.html
Availability details for
Quantity
  • Availability
  • Add to Cart