MAT047023670 Matrix Scientific

2'-Fluoro-3'-(trifluoromethyl)acetophenone

  • MDL number MFCD00061145
  • Cas Number : 207853-63-2
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.02
Country of Origin USA
Product Link http://www.matrixscientific.com/001437.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001437.html
Availability details for
Quantity
  • Availability
  • Add to Cart