MAT047023679 Matrix Scientific

3,5-Dichlorobenzyl chloride

  • MDL number MFCD00060278
  • Cas Number : 3290-06-0
  • Molecular Weight : 195.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36
Purity 98
Country of Origin USA
Boiling point 115-123� Torr
Product Link http://www.matrixscientific.com/005240.html

Safety Information

hcodes Corrosive/Lachrymator
MSDS http://www.matrixscientific.com/005240.html
Availability details for
Quantity
  • Availability
  • Add to Cart