MAT047023681 Matrix Scientific

4'-Fluoro-3'-(trifluoromethyl)acetophenone

  • MDL number MFCD00061252
  • Cas Number : 208173-24-4
  • Molecular Weight : 206.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001438.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001438.html
Availability details for
Quantity
  • Availability
  • Add to Cart