MAT047023683 Matrix Scientific

2,4-Difluorobenzyl alcohol

  • MDL number MFCD00009983
  • Cas Number : 56456-47-4
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.29
Country of Origin USA
Boiling point 52�m
Product Link http://www.matrixscientific.com/005243.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005243.html
Availability details for
Quantity
  • Availability
  • Add to Cart