MAT047023710 Matrix Scientific

1-[4-(2-Chloroethyl)phenyl]-1-ethanone

  • MDL number MFCD00016350
  • Cas Number : 69614-95-5
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.15
Storage temp. Ambient
Country of Origin USA
Boiling point 112�1 Torr
Product Link http://www.matrixscientific.com/005274.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005274.html
Availability details for
Quantity
  • Availability
  • Add to Cart