MAT047023734 Matrix Scientific

2,6-Difluoromandelic acid

  • MDL number MFCD00061298
  • Cas Number : 207981-50-8
  • Molecular Weight : 188.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005298.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005298.html
Availability details for
Quantity
  • Availability
  • Add to Cart