MAT047023737 Matrix Scientific

2-Hydroxy-3-(trifluoromethyl)pyridine

  • MDL number MFCD00153194
  • Cas Number : 22245-83-6
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148�ubl)
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001443.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001443.html
Availability details for
Quantity
  • Availability
  • Add to Cart