MAT047023740 Matrix Scientific

3-(Trifluoromethoxy)benzonitrile

  • MDL number MFCD00040959
  • Cas Number : 52771-22-9
  • Molecular Weight : 187.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005303.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/005303.html
Availability details for
Quantity
  • Availability
  • Add to Cart