MAT047023751 Matrix Scientific

2-(Trifluoromethoxy)benzamide

  • MDL number MFCD00042407
  • Cas Number : 127979-74-2
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-155
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005313.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005313.html
Availability details for
Quantity
  • Availability
  • Add to Cart