MAT047023756 Matrix Scientific

3-(Trifluoromethoxy)benzylamine

  • MDL number MFCD00061267
  • Cas Number : 93071-75-1
  • Molecular Weight : 191.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005319.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/005319.html
Availability details for
Quantity
  • Availability
  • Add to Cart