MAT047023758 Matrix Scientific

3-(Trifluoromethoxy)phenylacetonitrile

  • MDL number MFCD00061272
  • Cas Number : 108307-56-8
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005321.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/005321.html
Availability details for
Quantity
  • Availability
  • Add to Cart