MAT047023763 Matrix Scientific

2-(Trifluoromethoxy)fluorobenzene

  • MDL number MFCD00236322
  • Cas Number : 2106-18-5
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 105-106
Product Link http://www.matrixscientific.com/005327.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/005327.html
Availability details for
Quantity
  • Availability
  • Add to Cart