MAT047023765 Matrix Scientific

2-(Trifluoromethoxy)phenylacetic acid

  • MDL number MFCD00236325
  • Cas Number : 220239-67-8
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-56
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/005329.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005329.html
Availability details for
Quantity
  • Availability
  • Add to Cart