MAT047023775 Matrix Scientific

3-(Trifluoromethoxy)anisole

  • MDL number MFCD01631504
  • Cas Number : 142738-94-1
  • Molecular Weight : 192.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/005340.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005340.html
Availability details for
Quantity
  • Availability
  • Add to Cart