MAT047023879 Matrix Scientific

3-(Trifluoromethyl)phenyl acetate

  • MDL number MFCD01861206
  • Cas Number : 78950-34-2
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/005447.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005447.html
Availability details for
Quantity
  • Availability
  • Add to Cart