MAT047023882 Matrix Scientific

5-(Trifluoromethyl)benzo-[2,1,3]-thiadiazole

  • MDL number MFCD00114584
  • Cas Number : 17754-05-1
  • Molecular Weight : 204.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Storage temp. Ambient
Country of Origin USA
Boiling point 100�mm
Product Link http://www.matrixscientific.com/005454.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005454.html
Availability details for
Quantity
  • Availability
  • Add to Cart