MAT047023902 Matrix Scientific

2-(Trifluoromethyl)-2-hydroxypropionic acid

  • MDL number MFCD00190646
  • Cas Number : 374-35-6
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88
Purity 95
Country of Origin USA
Boiling point 189
Product Link http://www.matrixscientific.com/005472.html

Safety Information

hcodes Corrosive/Irritant
MSDS http://www.matrixscientific.com/005472.html
Availability details for
Quantity
  • Availability
  • Add to Cart