MAT047023910 Matrix Scientific

4-Methoxy-3-methylphenylacetic acid

  • MDL number MFCD00041034
  • Cas Number : 4513-73-9
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001462.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001462.html
Availability details for
Quantity
  • Availability
  • Add to Cart