MAT047023942 Matrix Scientific

2-(Trifluoromethyl)phenyl isothiocyanate

  • MDL number MFCD00039644
  • Cas Number : 1743-86-8
  • Molecular Weight : 203.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.346
Country of Origin USA
Boiling point 96� Torr
Product Link http://www.matrixscientific.com/005514.html

Safety Information

hcodes Toxic/Corrosive/Lachrymator
MSDS http://www.matrixscientific.com/005514.html
Availability details for
Quantity
  • Availability
  • Add to Cart