MAT047023957 Matrix Scientific

4-(Trifluoromethylthio)phenylacetic acid

  • MDL number MFCD00236351
  • Cas Number : 102582-93-4
  • Molecular Weight : 236.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005535.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005535.html
Availability details for
Quantity
  • Availability
  • Add to Cart