MAT047023959 Matrix Scientific

3-(Trifluoromethylthio)phenylacetonitrile

  • MDL number MFCD00236353
  • Cas Number : 82174-09-2
  • Molecular Weight : 217.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/005537.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/005537.html
Availability details for
Quantity
  • Availability
  • Add to Cart