MAT047023977 Matrix Scientific

4-Methoxy-3-nitrobenzotrifluoride

  • MDL number MFCD00007099
  • Cas Number : 394-25-2
  • Molecular Weight : 221.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-49
Purity 98
Country of Origin USA
Boiling point 96-98�8mm
Product Link http://www.matrixscientific.com/001468.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001468.html
Availability details for
Quantity
  • Availability
  • Add to Cart