MAT047024028 Matrix Scientific

Dodecafluorosuberic acid

  • MDL number MFCD00153146
  • Cas Number : 678-45-5
  • Molecular Weight : 390.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-144
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/005614.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/005614.html
Availability details for
Quantity
  • Availability
  • Add to Cart