MAT047024032 Matrix Scientific

3'-Fluoro-4'-(trifluoromethyl)propiophenone

  • MDL number MFCD00236306
  • Cas Number : 237761-78-3
  • Molecular Weight : 220.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/001474.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001474.html
Availability details for
Quantity
  • Availability
  • Add to Cart