MAT047024041 Matrix Scientific

3,4-Difluorotoluene

  • MDL number MFCD00075087
  • Cas Number : 2927-34-6
  • Molecular Weight : 128.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.12
Flash Point 25
Country of Origin USA
Boiling point 112-113
Product Link http://www.matrixscientific.com/005628.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/005628.html
Availability details for
Quantity
  • Availability
  • Add to Cart