MAT047024054 Matrix Scientific

2-Bromo-3',4'-difluoroacetophenone

  • MDL number MFCD00085010
  • Cas Number : 40706-98-7
  • Molecular Weight : 235.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/001062.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/001062.html
Availability details for
Quantity
  • Availability
  • Add to Cart