MAT047024055 Matrix Scientific

3-Chloro-4-fluoroacetophenone

  • MDL number MFCD00042203
  • Cas Number : 2923-66-2
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Purity 97
Country of Origin USA
Boiling point 126-128�mm
Product Link http://www.matrixscientific.com/001477.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001477.html
Availability details for
Quantity
  • Availability
  • Add to Cart